Cole-Parmer® Essentials Slot Samplers, Sterile HDPE
Reduce cross contamination while sampling various powders, granules, and grains
  • Suitable for sampling from all layers of the container, providing an accurate representation of the contents
  • Constructed of chemically inert high-density polyethylene (HDPE)
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness. Samplers are also sterilized via gamma irradiation (25-45 kGy) and individually bagged to reduce contamination.
$472.22 - $663.76CAD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Volume (mL) Description
Availability Pricing
COMPARE
62 100 Slot Sampler, HDPE, Sterile, 3 Openings, 100 mL; 20/PK
$511.27CAD / Pkg of 20
COMPARE
97 170 Slot Sampler, HDPE, Sterile, 3 Openings, 170 mL; 20/PK
$663.76CAD / Pkg of 20
COMPARE
62 50 Slot Sampler, HDPE, Sterile, 1 Opening, 62 cm Length; 20/PK
$472.22CAD / Pkg of 20
COMPARE
97 50 Slot Sampler, HDPE, Sterile, 1 Opening, 97 cm Length; 20/PK
$590.09CAD / Pkg of 20