Cole-Parmer® Essentials Pharma Lab Sampling Scoops, 316L Stainless Steel
Polished smooth to ensure materials cannot hide in crevices
  • Heavy gauge 316L stainless steel offers excellent corrosion resistance and strength
  • Ideal for all types of work from the laboratory to production
  • BSE/TSE free, GMP compliant, and autoclavable
High-quality, 316L stainless steel lab scoops provide corrosion resistance, particularly against chlorides and other industrial solvents and feature a hygienic design ideal for the pharmaceutical and food industries. All welds are ground and polished to a high-quality mirror finish better than 0.5 microns Ra to ensure they are free of crevices where powders and granules can get stuck. This provides excellent protection from cross-contamination. The corners on these lab scoops are radiused making them easy to clean. These sampling scoops are also BSE/TSE free, GMP compliant, and autoclavable.
When used in place of disposable scoops, these reusable lab scoops help reduce waste in the laboratory, as well as save you money and reordering time. Indicated capacities are approximate and are not intended for measuring specific volumes.
$140.12 - $436.86CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Diameter (cm) Overall Length (cm)
Availability Pricing
COMPARE
10 2.5 7.5
$140.12CAD / Each
COMPARE
50 3.8 11
$168.33CAD / Each
COMPARE
100 5 18
$196.33CAD / Each
COMPARE
250 7.5 23.5
$217.69CAD / Each
COMPARE
500 9.5 24
$259.32CAD / Each
COMPARE
750 9.5 33
$302.41CAD / Each
COMPARE
1000 10 34
$307.94CAD / Each
COMPARE
1250 12.5 30.5
$335.93CAD / Each
COMPARE
1700 13.5 35.5
$324.88CAD / Each
COMPARE
2500 18.5 32
$434.28CAD / Each
COMPARE
5000 22 35.5
$436.86CAD / Each
Showing 1- 11 of 11