Cole-Parmer® Essentials Multichannel Pipettor Racks
Unique design accommodates both multichannel and standard pipettes
  • The patented open slot allow multichannel head to fit on the rack
  • 5-mm thick acrylic offers durability along with stability
  • Four nonskid rubber feet provide extra stability
Store single-channel pipettors along with multichannel pipettors on a single pipette rack. A unique design featuring a slot accommodates a multichannel head longways through the rack. Pipettors rest securely in place at two places, under the finger hook and near the bottom of the handle, for added stability.
$88.40 - $140.71CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With
Availability Pricing
COMPARE
Acrylic 3 pipettors, up to 1 multichannel pipettor
$88.40CAD / Each
COMPARE
Acrylic 5 pipettors, up to 2 multichannel pipettors
$119.79CAD / Each
COMPARE
Acrylic 6 pipettors, up to 4 multichannel pipettors
$132.31CAD / Each
COMPARE
Acrylic 8 pipettors, up to 6 multichannel pipettors
$140.71CAD / Each