Cole-Parmer® Essentials Liquid Sampler Collectors, HDPE
Reduce cross contamination while sampling low viscosity liquids and oils
  • Manufactured in Class 100,000 cleanroom and individually packaged
  • Constructed of chemically inert high-density polyethylene (HDPE)
The liquid sampler collectors are designed to easily take liquid samples with the added convenience of not having to clean the sampler prior to use. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness and individually bagged. Choose from sterile or non-sterile samplers. Sterile samplers are sterilized via gamma irradiation (25k Gy minimum).
$330.41 - $523.05CAD / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (cm) Sterile Volume (mL)
Availability Pricing
COMPARE
50 No 100
$330.41CAD / Pkg of 20
COMPARE
50 Yes 100
$395.97CAD / Pkg of 20
COMPARE
100 No 190
$452.33CAD / Pkg of 20
COMPARE
100 Yes 190
$523.05CAD / Pkg of 20