Cole-Parmer® Essentials Digital Counter Pens
Combination marker and digital counter reduces errors—even when interrupted!
  • Automatically beeps every time a touch is made
  • Audio and visual counts to 99,999
  • Easy-to-read LCD
  • Felt-tipped pen with black water based ink is not permanent
Counter-Pen is a combination marker and digital counter designed for the lab and plant. Use it to count cell growth on petri dishes, inventory parts, and tally samples received. Counter beeps each time a mark is made ensuring an accurate tally. LCD simultaneously displays total and can be reset at the touch of a button. Durable ABS plastic pen features on/off switch and comes with cap.
$37.57CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Battery Material Description
Availability Pricing
COMPARE
1 392 Silver Oxide (included) ABS Plastic Digital Counter Pen
$37.57CAD / Each