Cole-Parmer Epoxy-Body Special-purpose Electrodes
Unique probes for special applications
  • Combine quality with durability
  • Eposy-body housing resists impact
  • Compatible with most pH meters
These quality electrodes have a no maintenance sealed gel reference. All electrodes have a Ag/AgCl reference cell.
$218.06 - $302.78CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Body Material Electrode Type Reference Junction
Availability Pricing
COMPARE
Epoxy Single junction PTFE
$302.78CAD / Each
COMPARE
Epoxy Double junction
$218.06CAD / Each