This product is now discontinued, View Related Products
Cole-Parmer® Electronic Multichannel Pipettors
Multiple modes meet your pipetting needs
  • Preprogrammed with six different operations
  • Adjustable aspiration and pipetting speeds
  • Large color LED display is easy to read
  • Lightweight, ergonomic, and simple to use
The electronic multichannel pipettors feature a large easy-to-read color LED glass display and a simple to use menu. Multiple modes of pipetting are available: forward, gel loading, manual, mixing, reverse, sample mixing, sub-sample, titration, and unequal volume.
You can program the pipettor with your own protocol or choose from one of the six preprogrammed operations. The pipettors can be used with a variety of liquids including easy-foaming, high-viscosity, and volatile liquids. Other features include soft tip ejection, internal automatic calibration, adjustable aspiration, and autoclavable tip cone. The entire pipettor can also be sterilized by UV light.
Choose from 8- or 12-channel pipettors to meet your laboratory needs.
$1,735.65 - $1,880.78CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accuracy (%) Max Volume (µL) Number Of Channels
Availability Pricing
COMPARE
±2.5 10 8
$1,735.65CAD / Each
COMPARE
±2.0 100 8
$1,735.65CAD / Each
COMPARE
±2.5 300 8
$1,759.23CAD / Each
COMPARE
±2.5 10 12
$1,880.78CAD / Each
COMPARE
±2.0 100 12
$1,880.78CAD / Each
COMPARE
±2.5 300 12