This product is now discontinued, View Related Products
Cole-Parmer Economy Lab pH Meters
Designed for lab use
  • Display is ideal for quick checks at a glance
  • High accuracy at an exceptional price
Lab meters are ideal for benchtop use—feature solid construction and economical design. They're accurate to ±0.01 pH unit (±1 mV). Lab meters accept electrodes with BNC connectors (order separately). Meters operate on eight AA alkaline batteries (included), or optional 110 or 220 VAC adapters (order separately).
$659.19 - $694.85CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max mV (mV) Max pH (pH) Description
Availability Pricing
COMPARE
700 14 laboratory pH/mV meter, analog
$659.19CAD / Each
COMPARE
1999 14 laboratory pH/mV meter, digital
$694.85CAD / Each