Cole-Parmer Economy Compact Dual-Power Stereomicroscopes
Won’t take up a lot of space on your lab bench
  • Adjust head and eyepieces reduce eye strain
These compact dual-power stereomicroscopes stand 11'' tall have a working distance of 75 mm. They feature a 45° inclined swivel head with adjustable diopter and interpupillary controls. Heads rotate 360° for front position viewing. Paired objectives are mounted in a revolving turret. Paired 10x widefield eyepieces have a 34-mm OD and accept reticles with 22.8-mm OD. Use optional 15x eyepieces for greater magnifying power. Stereomicroscopes are supported by a cast metal frame with gray enamel finish.
Built-in incident (top) and transmitted (bottom) light sources on all models allow versatile illumination of any sample. Each stereomicroscope is supplied with both frosted glass and reversible black-and-white stage plates and a vinyl dust cover. Include a 6-ft cord with three-prong plug.
$481.80CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Magnification
Availability Pricing
COMPARE
10x/30x
$481.80CAD / Each