This product is now discontinued, View Related Products
Cole-Parmer Dual Vertical Mini-Gel Systems
Customize your own gel system depending on your needs and budget!
Mini Dual Gel Units. Small format is ideal for mini SDS-PAGE and mini agarose gel separations, plus it reduces the use of expensive reagents. These dual units let you run two agarose or polyacrylamide gels simultaneously under identical conditions; also compatible with most precast gels. The built-in cooling system ensures fast run times and flat, even banding patterns. Chamber gasket and clamps create a positive, leakproof seal.
Required System Components:
1) Gel Unit (order power supply separately)
For the Gel Wrap™ Gasket gel casting method, you will need:
2) Glass plate set(s)
3) Casting clamps (three per plate set for mini units)
Order the same thickness of:
4) GelWrap gasket (one per plate set)
5) Spacer set (one per plate set)
6) Combs (one per plate set)
$842.78 - $872.25CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Glass Height (cm) Glass Width (cm)
Availability Pricing
COMPARE
250 10 11.3
$872.25CAD / Each
COMPARE
250 8 10
$842.78CAD / Each