This product is now discontinued, View Related Products
Cole-Parmer® Drum Spill Pallet and Ramp
Protect your plant from messy drum leaks
  • Load drums in a cinch with the durable spill pallet ramp
  • Tough, one-piece polyethylene construction will not rust or corrode
  • Reliably catch incidental leaks with large spill capacity
  • Low-profile spill pallet optimizes space and efficiency while enhancing safety
  • Easily clean with removable heavy-duty structural foam grates
This Cole-Parmer® low-profile drum spill pallet is made of an all polyethylene construction to meet a wide range of chemical compatibility needs. The spill pallet collects small leaks and spills to create a safer workspace.
The work ramp allows for easy loading of drums into the low-profile spill pallet to provide maximum efficiency with minimal effort. The work ramp is small enough to stay out of your way, saving you valuable work and floorspace. Improved non-skid surface eliminates risk of slipping and 10° slope provides easy loading. Purchase ramp and pallet separately.
$312.36 - $331.51CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Height (in) Material Description
Availability Pricing
COMPARE
25 1/2 Polyethylene Drum Ramp for Low Profile Spill Pallets
$331.51CAD / Each
COMPARE
28 1/2 Polyethylene Low-Profile Drum Spill Pallet, 2-Drum; 22 gal
$312.36CAD / Each