This product is now discontinued, View Related Products
Cole-Parmer® Drum Spill Decks
Protect your plant from messy drum leaks
  • Modular, locking design allows for multiple drum customization
  • Tough, one-piece polyethylene construction will not rust or corrode
  • Reliably catch incidental leaks with large spill capacity
These Cole-Parmer® drum spill decks are made of an all polyethylene construction to meet a wide range of chemical compatibility needs. Spill decks collect small leaks and spills to create a safer workspace. The modular, locking design enables you to combine more than one drum deck to a create a corner unit or 4-drum unit. The drum deck is light-weight and can be easily be moved to different areas within your plant while giving you the reliability to handle your heaviest drums.
$142.92 - $179.75CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Height (in) Description
Availability Pricing
COMPARE
17 29 1/2 Drum Spill Deck, 1-Drum; 17 gal
$142.92CAD / Each
COMPARE
33 57 Drum Spill Deck, 2-Drum; 33 gal
$179.75CAD / Each