Cole-Parmer® Essentials Inoculating Loop and Needles, Sterile, Polystyrene
Sterile inoculating loops with needles save time and eliminate cross-contamination when transferring solid and liquid media
  • Loop and needle combination for dual function
  • Use straight from the pack without sterilizing
  • Flexible and disposable
  • Treated, hydrophilic surface
A loop on one end and needle on the other make these disposable inoculating lab tools suitable for both solid and liquid media. They are vacuum-gas plasma surface treated to allow culture medium to attach. The long, flexible stem of these inoculating loops can be bent allowing access to narrow or small-volume tubes and dishes. Color coding identifies inoculation volume easily.
Manufactured under ISO 13485 and ISO 9001 quality management systems in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these radiation sterilized (SAL 10-6) inoculating loops are available in individual peel-to-open packs, or in resealable packs of 10 or 25. Every package is printed with a lot number for quality traceability. DNase/RNase free and nonpyrogenic.
$60.41 - $162.07CAD / Case of 1000
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Color Contained Volume (µL) Overall Length (mm)
Availability Pricing
COMPARE
Blue 1 228
$120.82CAD / Case of 1000
COMPARE
Blue 1 228
$162.07CAD / Case of 1000
COMPARE
Yellow 10 228
$120.82CAD / Case of 1000
COMPARE
Yellow 10 228
$162.07CAD / Case of 1000
COMPARE
Blue 1 228
$60.41CAD / Case of 250
COMPARE
Yellow 10 228
$60.41CAD / Case of 250