Cole-Parmer® RH-800 Series Digital Plus Heated Circulating Baths
Keep temperature steady at point of use
  • Closed loop functionality provides greater temperature stability at the external apparatus
  • Temperature calibration increases accuracy
  • Intuitive interface uses readily recognizable icons
  • Controller panel swivels for easy viewing from any angle
These integrated heated circulators offer reliable, stable temperature control. Single-point temperature calibration allows you to program an offset temperature, enhancing temperature control precision. Large, backlit LCD shows set point and actual bath temperatures simultaneously. Safety and convenience features include overtemperature protection, failsafe heater control, high and low temperature limits with alarms, low-liquid level alarm, alarm and fault messages, and automatic loss of power reset.
$4,717.79 - $6,017.32CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Power (VAC) Description
Availability Pricing
COMPARE
200 115 Digital Plus Heated Circulator, 7L; 120V/60Hz
$4,981.53CAD / Each
COMPARE
200 230 Digital Plus Heated Circulator, 7L; 240V/50Hz
$4,981.53CAD / Each
COMPARE
200 115 Digital Plus Heated Circulator, 15L; 120V/60Hz
$5,569.41CAD / Each
COMPARE
200 230 Digital Plus Heated Circulator, 15L; 240V/50Hz
$4,717.79CAD / Each
COMPARE
200 115 Digital Plus Heated Circulator, 20L; 120V/60Hz
$6,017.32CAD / Each
COMPARE
200 230 Digital Plus Heated Circulator, 20L; 240V/50Hz
$6,017.32CAD / Each