This product is now discontinued, View Related Products
Cole-Parmer Digital Gauge with Visual Alarm
All stainless steel sensor is vibration, shock, and weather resistant
  • Tare, peak, and hold functions are standard
  • Auto-off after 5 minutes
  • User-selectable units of measure; PSI, BAR, MPA
$748.11 - $1,053.47CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±0.25% full-scale 0.1\" Hg
$1,053.47CAD / Each
COMPARE
±0.25% full-scale 15 0.01 psi
$750.69CAD / Each
COMPARE
±0.25% full-scale 30 0.01 psi
$1,035.79CAD / Each
COMPARE
±0.25% full-scale 60 0.01 psi
$748.11CAD / Each
COMPARE
±0.25% full-scale 100 0.1 psi
$768.00CAD / Each
COMPARE
±0.25% full-scale 500 0.1 psi
$768.00CAD / Each
COMPARE
±0.25% full-scale 1000 1 psi
$768.00CAD / Each
COMPARE
±0.25% full-scale 4000 1 psi
$927.50CAD / Each
COMPARE
±0.25% full-scale 9999 1 psi
$768.00CAD / Each
Showing 1- 9 of 9