This product is now discontinued, View Related Products
Foxx Life Sciences Die-Cut Filter Membranes
Economical, efficient surface filtering for the retention of particles or microorganisms
  • Materials adhere to USP Class VI Standards and FDA compliances
Nylon membranes offer good chemical resistance, making them ideal for filtering your more aggressive solutions such as alcohols and DMSO solutions.
Hydrophobic PTFE membranes are bonded to a polypropylene support for improved handling. They are highly compatible with almost all solvents including strong acids and bases.
Mixed cellulose ester (MCE) membranes are gridded for easy cell counts. These sterile disc filters are used for the microbiological analysis of water and other fluids in the food & beverage and pharmaceutical industries. Sterilized by gamma irradiation and individually packaged.
$35.73 - $287.31CAD / Pkg of 100
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Material Pore Size (µm)
Availability Pricing
COMPARE
13 PES 0.22
$213.64CAD / Pkg of 100
COMPARE
25 PES 0.22
$235.74CAD / Pkg of 50
COMPARE
47 PES 0.22
$279.94CAD / Pkg of 50
COMPARE
90 PES 0.22
$114.92CAD / Pkg of 25
COMPARE
13 PES 0.45
$213.64CAD / Pkg of 100
COMPARE
25 PES 0.45
$243.11CAD / Pkg of 50
COMPARE
47 PES 0.45
$287.31CAD / Pkg of 50
COMPARE
90 PES 0.45
$114.92CAD / Pkg of 25
COMPARE
25 Nylon 0.22
$35.73CAD / Pkg of 50
COMPARE
47 Nylon 0.22
$45.31CAD / Pkg of 50
COMPARE
90 Nylon 0.22
$65.93CAD / Pkg of 25
COMPARE
25 Nylon 0.45
$35.73CAD / Pkg of 50
Foxx Life Sciences Filter Membrane, 47 mm, .45 µm, nylon; 50/pk RK-06060-16 Mfr # 364-3612-OEM
COMPARE
47 Nylon 0.45
$51.57CAD / Pkg of 50
COMPARE
90 Nylon 0.45
$51.94CAD / Pkg of 25
COMPARE
25 Hydrophobic PTFE 0.22
$51.94CAD / Pkg of 50
COMPARE
47 Hydrophobic PTFE 0.22
$73.67CAD / Pkg of 50
COMPARE
90 Hydrophobic PTFE 0.22
$145.87CAD / Pkg of 25
COMPARE
25 Hydrophobic PTFE 0.45
$52.67CAD / Pkg of 50
COMPARE
47 Hydrophobic PTFE 0.45
$73.67CAD / Pkg of 50
COMPARE
90 Hydrophobic PTFE 0.45
$190.07CAD / Pkg of 25
Showing 1 - 20 of 24