This product is now discontinued, View Related Products
Cole-Parmer Desi-Vac Vaccum Desiccators
Built-in pump achieves a vacuum of >200" H2O
Self-contained polycarbonate containers are ideal for drying, preserving, storing, and shipping samples. Built-in vacuum pump removes air via manual strokes. Rubber O-ring ensures a tight seal and maintains a vacuum of >200" H2O (14.7" Hg) for over 24 hours. Quick-release button allows air back into unit to remove the lid.
Large capacity model 06525-09 features an automatic vacuum pump that removes air and maintains a countinuous vacuum. This unit also includes a sample of desiccant drying packets.
$41.99 - $135.55CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Chamber Volume (Liters)
Availability Pricing
COMPARE
0.7
$53.04CAD / Each
COMPARE
0.85
$41.99CAD / Each
COMPARE
1.5
$68.51CAD / Each
COMPARE
3
$69.25CAD / Each
COMPARE
12
$135.55CAD / Each