This product is now discontinued, View Related Products
Cole-Parmer Data Logging Digital Gauges with Bar Graph Display
270° bar graph reads 0 to 100% of scale
  • Log up to 60 records — time interval or event triggered
  • Features tare, peak, hold, and min/max functions
  • Selectable auto-off of 2, 4, 8, 16, and 32 minutes
Gauges feature user-selectable units of measure and a built-in lockout feature that prevents unauthorized access. Menu is easily accessed via the four-button controls on face of gauge. Use in many applications such as line pressure, vessel pressure, and force measurement. Large, 5-1/2" display allows you to read pressure via bar graph or numeric readout at far distances.
Choose the battery-operated units for remote monitoring of pressure where power is not accessible, or choose the voltage-powered units with 4 to 20 mA output. Units with 4 to 20 mA output allow you to send pressure measurement readings to recorders, data loggers, or PLCs for switching or pump control applications.
$1,008.90 - $1,387.20CAD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 0.01
$1,131.20CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 15 0.01
$1,019.59CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 30 0.01
$1,008.90CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 60 0.01
$1,028.79CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 100 0.01
$1,019.59CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 500 0.1
$1,019.59CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 1000 1
$1,019.59CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 5000 1
$1,102.10CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 10000 1
$1,019.59CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 0.01
$1,387.20CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 15 0.01
$1,218.86CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 30 0.01
$1,218.86CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 60 0.01
$1,218.86CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 100 0.01
$1,218.86CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 500 0.1
$1,218.86CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 1000 1
$1,218.86CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 5000 1
$1,387.20CAD / Each
COMPARE
±0.25% full-scale (BFSL) or two digit counts, whichever is greater 1
$1,218.86CAD / Each
Showing 1- 18 of 18