This product is now discontinued, View Related Products
Cole-Parmer® Essentials Dark Amber Carboys, Polypropylene
Store light sensitive solutions
  • Maximize storage efficiency with rectangular shape
  • Secure grips make lifting, handling, and pouring easy
  • Large metric graduations are certified to ±5% accuracy
  • USP Class VI, FDA-compliant, BPA-free, autoclavable material
Recommended autoclaving cycle is 15 PSI for 20 minutes at 121° C, with cap off or placed on top (not screwed in place).
$223.96 - $381.61CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Description
Availability Pricing
COMPARE
20 Dark Amber PP Carboy, 83B Cap, 20 L; 1/Ea
$223.96CAD / Each
COMPARE
20 Dark Amber PP Carboy, 120 mm VersaCap, 20 L; 1/Ea
$241.64CAD / Each
COMPARE
40 Dark Amber PP Carboy, 120 mm VersaCap, 40L; 1/Ea
$381.61CAD / Each