This product is now discontinued, View Related Products
Cole-Parmer® Essentials Class A Graduated Glass Cylinders - To Contain
Heavy hexagonal base provides stability to prevent cylinder from tipping over
  • Class A borosilicate cylinders are calibrated "to contain"
  • Withstand temperatures up to 752°F (400°C) for short-term exposure and 445°F (230°C) for long-term exposure
  • Graduations are durable white enamel
  • Reference: ASTM E1272 Class A tolerances and specifications
  • Autoclavable
$33.15 - $72.20CAD / Pkg of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
COMPARE
50 1 Glass
$33.15CAD / Pkg of 1
COMPARE
100 1 Glass
$37.57CAD / Pkg of 1
COMPARE
250 2 Glass
$59.89CAD / Pkg of 1
COMPARE
500 5 Glass
$72.20CAD / Pkg of 1