This product is now discontinued, View Related Products
Cole-Parmer Class A Glass Erlenmeyer Flasks
Low-cost economical alternative fits every budget!
  • Narrow-mouth opening with beaded heavy-duty rim
  • Approximate graduations and marking spot are durable white enamel
  • Withstands short-term exposure to 752°F (400°C) and long-term exposure to 445°F (230°C)
  • Class A borosilicate glass meets ASTM E 1404, DIN 12380 specifications
  • Autoclavable
$43.54 - $90.10CAD / Pkg of 12
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Stopper Number Accepted
Availability Pricing
COMPARE
100 25 5
$61.44CAD / Pkg of 12
COMPARE
250 25 6
$61.44CAD / Pkg of 12
COMPARE
500 100 7
$43.54CAD / Pkg of 6
COMPARE
1000 50 9
$70.72CAD / Pkg of 6
COMPARE
2000 100 10
$52.89CAD / Pkg of 1
COMPARE
3000 500 10
$90.10CAD / Pkg of 1