This product is now discontinued, View Related Products
Cole-Parmer Class A Burettes (Burets)
Positive threaded-back PTFE stopcock won't come loose during titrations
  • Class A burettes (burets) with clear borosilicate bodies and chemical-resistant PTFE stopcocks
  • White graduation markings are easy to read
These Class A burettes (burets) are calibrated at 20°C to exceed ASTM E287 (meets or exceeds DIN, ISO, and ASTM standards). They feature a maximum temperature of 460°F (240°C) for normal use and 750°F (400°C) for short-term use.
$138.42 - $191.98CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Tolerance
Availability Pricing
COMPARE
10 0.05 0.02
$138.42CAD / Each
COMPARE
25 0.1 0.03
$147.34CAD / Each
COMPARE
50 0.1 0.05
$150.43CAD / Each
COMPARE
100 0.2 0.1
$191.98CAD / Each