Cole-Parmer Calcium Chloride Salt Glass Hydrometers (Salometers)
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
Ideal for determining the percentage of saturation, specific gravity or freezing point of CaCl2. Each hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
$59.67 - $61.00CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0/120 Percent Saturation Calcium Chloride Salometer
$59.67CAD / Each
COMPARE
Sp.Gr. 1.000/1.280 Calcium Chloride Salometer
$61.00CAD / Each