This product is now discontinued, View Related Products
Cole-Parmer Brass Case Indicators
Provide fast, accurate measurements
Choose an indicator for %RH measurement only, %RH and °>F measurements, or %RH, °>F and °>C measurements. All models have a cellulose RH sensor that measures relative humidity from 0 to 100% RH. Thermohygrometers feature a bimetallic temperature sensor.
Models 03310-20 and 03310-40 include a test certificate ensuring conformance to stated specifications. All indicators have a solid brass case, a 5" diameter glass face, and predrilled holes for wall mounting.
$262.26 - $499.11CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max RH Range (%) Max Temperature (° F) Min RH Range (%)
Availability Pricing
COMPARE
100 170 0
$262.26CAD / Each
COMPARE
100 170 0
$440.54CAD / Each
COMPARE
100 170 0
$499.11CAD / Each