Cole-Parmer® ATC/Grounded Housings
Feature built-in ATC element and solution ground
  • A variety of housing styles available to meet your application requirements
  • Maximum pressure is 100 psi
These unique holders feature an integrated 316 stainless steel pin ground wire that serves as a temperature compensator and a solution ground. Choose economical CPVC housing or chemical-resistant KYNAR housing. In-line holders have a 1'' NPT(M) connection. Submersible holders have a 3/4'' NPT(F) thread--connect plastic pipe to match your required submersion depth.
$244.58CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Electrode Housing, submersible, CPVC, no ATC
$244.58CAD / Each