Cole-Parmer® Essentials Analytical Funnel, PTFE
Withstands most chemical compounds
  • Chemically inert
  • Autoclavable
Suitable for a variety of laboratory applications, these high purity funnel feature non-stick internal finish. Funnel can withstand most chemical compounds and corrosive environments. Temperature usage is 0 to 280°C (32 to 536°F).
$90.61 - $691.76CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Stem OD (mm) Top OD (mm)
Availability Pricing
COMPARE
5 8 30
$90.61CAD / Each
COMPARE
23 10 50
$124.50CAD / Each
COMPARE
70 12 70
$186.38CAD / Each
COMPARE
195 14 100
$362.45CAD / Each
COMPARE
700 16 150
$691.76CAD / Each