This product is now discontinued, View Related Products
Cole-Parmer Analog Melting Point Apparatuses
Safe, simple to use, and accurate
  • Easily view the melt via magnifying lens with built-in illumination
  • Be more productive—test multiple samples simultaneously
The analog melting point apparatus features a blue-spirit-filled thermometer for mercury-free temperature measurements. The unit is simple to operate with instructions printed clearly on the front panel — ideal for educational settings. Test and view up to three samples at once for greater efficiency. The ramp rate is 1 to 20°C per minute until melt occurs.
$1,380.57 - $1,561.79CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Heat-Up Time Max Temperature (° C) Power (VAC)
Availability Pricing
COMPARE
50 to 350⁰C in 15 minutes 250 120
$1,380.57CAD / Each
COMPARE
50 to 350⁰C in 15 minutes 250 230
$1,561.79CAD / Each