Cole-Parmer Analog Controlled Heating Mantles
Heat samples safely, efficiently, and faster
  • Powder-coated aluminum case is cool to the touch and easy to clean
  • Heats to maximum temperature of 842°F (450°C)
Featuring a built-in power controller with a 1 to 10 scale dial, the electric heating mantles quickly heat liquid samples contained in the round-bottom flasks. A heating element suspended within a ceramic fiber insulating material provides safe heat transfer. Two neon lights indicate “power on” and “heat on.” The mantles handle a wide capacity of round bottom flasks between 50 and 1000 mL.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC)
Availability Pricing
COMPARE
50 120
Call For Price
COMPARE
50 230
Call For Price
COMPARE
100 120
Call For Price
COMPARE
100 230
Call For Price
COMPARE
250 120
Call For Price
COMPARE
250 230
Call For Price
COMPARE
500 120
Call For Price
COMPARE
500 230
Call For Price
COMPARE
1000 120
Call For Price
COMPARE
1000 230
Call For Price
Showing 1- 10 of 10