This product is now discontinued, View Related Products
Cole-Parmer Adjustable Height Single and Dual Vertical Gel Systems
Customize your own gel system depending on your needs and budget!
Versatile units are adjustable for different pathlengths. Ideal for SDS page, sequencing, mobility shift assays, and characterizing or separating proteins. The upper reservoir is height adjustable along the guide rods, and creates a positive, leakproof seal when plate assembly is clamped into place.
Required System Components:
1) Gel Unit (order power supply separately)
For the Gel Wrap™ Gasket gel casting method, you will need:
2) Glass plate set(s)
3) Casting clamps (six per plate set for height-adjustable units)
Order the same thickness of:
4) GelWrap gasket (one per plate set)
5) Spacer set (one per plate set)
6) Combs (one per plate set)
$1,149.24 - $1,523.49CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Glass Height (cm) Glass Width (cm)
Availability Pricing
COMPARE
725 16.5
$1,149.24CAD / Each
COMPARE
500 16.5
$1,243.54CAD / Each
COMPARE
825 16.5
$1,523.49CAD / Each
COMPARE
900 16.5
$1,464.55CAD / Each