Cole-Parmer® 100-Ohm Pt RTD Conductivity/Resistivity Cells
Economical replacements for a variety of controllers
  • Ten-foot cables with stripped ends
  • Flow-through, in-line, swage, and dip styles available
Cells withstand pressures up to 150 psi (at 135°C for SS; 25°C for CPVC and PVDF).
$221.75 - $515.69CAD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Conductivity Cell; K=0.1, SS, Swage; 100 Ohm Pt RTD ATC
$415.13CAD / Each
COMPARE
Conductivity Cell; K=1.0, SS, Swage; 100 Ohm Pt RTD ATC
$392.29CAD / Each
COMPARE
Conductivity Cell; K=0.01, SS, In-Line; 100 Ohm Pt RTD ATC
$428.02CAD / Each
COMPARE
Conductivity Cell; K=0.1, SS, In-Line; 100 Ohm Pt RTD ATC
$382.34CAD / Each
COMPARE
Conductivity Cell; K=1.0, SS, In-Line; 100 Ohm Pt RTD ATC
$355.09CAD / Each
COMPARE
Conductivity Cell; K=10.0 CPVC, In-Line; 100 Ohm Pt RTD ATC
$383.08CAD / Each
COMPARE
Conductivity Cell; K=10.0; Kynar, In-Line; 100 Ohm Pt RTD ATC
$515.69CAD / Each
COMPARE
Conductivity Cell; K=1.0; Epoxy, In-Line; 100 Ohm Pt RTD ATC
$341.83CAD / Each
COMPARE
Conductivity Cell; K=0.1; Glass, Dip style, 100 Ohm Pt RTD ATC
$325.99CAD / Each
COMPARE
Conductivity Cell; K=1.0 Kynar, In-Line; 100 Ohm Pt RTD ATC
$387.87CAD / Each
COMPARE
Conductivity Cell; K=1.0; Glass, Dip-Style, 100 Ohm Pt RTD ATC
$279.94CAD / Each
COMPARE
Conductivity Cell; K=1.0; Glass, Flow Through; 100 Ohm Pt RTD ATC
$492.85CAD / Each
COMPARE
Conductivity Cell; K=0.1; Epoxy, In-line; 100 Ohm Pt RTD ATC
$373.50CAD / Each
COMPARE
Conductivity Cell; K=10; Epoxy, In-line; 100 Ohm Pt RTD ATC
$406.66CAD / Each
COMPARE
Conductivity Cell, K=1.0, Epoxy, Dip-Style, 100 Ohm Pt RTD ATC
$221.75CAD / Each
Showing 1- 15 of 15