This product is now discontinued, View Related Products
Clear Drying Chambers
Use acrylic chambers with aluminum tops to accommodate three-shelf sample stands (order separately) for bulk freeze drying process with visual monitoring. Select model 03337-66 for up to eight additional valved ports (e.g. for Fast-Freeze Flasks). Three-shelf stands are independently height-adjustable. Available for ambient temperature use (vials, serum bottles, and bulk samples) or heated (to 40° C, variable temperature controlled) to accelerate freeze drying of bulk samples.
$2,534.23 - $3,388.80CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Power (VAC) Sublimation Rate (Liters/day)
Availability Pricing
Labconco 7442900 Clear Freeze Drying Chamber without Ports RK-03337-64 Mfr # 7442900
COMPARE
110 / 220
$2,534.23CAD / Each
Labconco 7443500 Clear Freeze Drying Chamber with 8 Ports RK-03337-66 Mfr # 7443500
COMPARE
4.5 115 2
$3,388.80CAD / Each