Polypropylene Burette Clamps
Soft rubber pads hold burette securely without scratching
  • Fits standard 12.5 mm (1/2") support rod
  • Chemical resistant
  • Autoclavable
The non-corroding polypropylene (PP) clamps hold burettes in place without covering graduations. Clamps include 3/8" and 1/2" bushings to fit any standard support rod.
$33.89 - $50.54CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accepts Rod Diameter (in) Clamp Material Description
Availability Pricing
Bel-Art 18204 Single-Style Burette Clamp RK-06703-10 Mfr # 182040000
COMPARE
0.375 Polypropylene (PP) Single-Style Burette Clamp
$33.89CAD / Each
Bel-Art 18206 Double -Style Burette Clamp RK-06703-12 Mfr # 182060000
COMPARE
0.375 Polypropylene (PP) Double -Style Burette Clamp
$50.54CAD / Each