Chemsteel Gear Pumps with Mechanical Seals
Self-priming and bidirectional capabilities offer excellent dosing, metering, and transfer of polymers and corrosive chemicals
  • Wear-resistant helical gears offer reduced noise levels of 10 dB
  • Heavy-duty three-piece design combines bearings and wear plates to easily service pump in the field
316 SS Gear Pumps with mechanical seals provide a cost-effective option for pumping a variety of fluids.
All pumps are close-coupled to standard 56C-face motors. Motor type is TEFC, totally enclosed, fan-cooled. The 115/230 VAC models are single phase. Three phase motors for variable-speed use with controllers are available. Call our Application Specialists to order.
Also contact our Application Specialists if your maximum viscosity requirements are higher.
Service kits include: drive and idler gears mounted to the shafts, keys, bearings, O-rings, and seal assemblies.
$3,394.69 - $9,798.04CAD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Differential Pressure (PSI) Max Flow Rate (GPM) Wetted Parts
Availability Pricing
Chemsteel S2036BCB-C1F50 PPS gear pump, 1.5 GPM, 115/230 VAC RK-70736-00 Mfr # OBS2036BCB-C1F50
COMPARE
100 1.5 PTFE-encapsulated silicone
$3,406.48CAD / Each
Chemsteel RM10216CW-F50 PPS gear pump, 0.58 GPM, 115/230 VAC RK-70736-03 Mfr # OBRM10216CW-F50
COMPARE
100 0.5 PTFE-encapsulated silicone
$7,211.51CAD / Each
Chemsteel S2046BCB-C1J47 PPS gear pump, 2.0 GPM, 115/230 VAC RK-70736-05 Mfr # OBS2046BCB-C1J47
COMPARE
100 2 PTFE-encapsulated silicone
$3,394.69CAD / Each
Chemsteel S2076BCB-C1M47 PPS gear pump, 4.0 GPM, 115/230 VAC RK-70736-15 Mfr # OBS2076BCB-C1M47
COMPARE
100 4 PTFE-encapsulated silicone
$3,901.54CAD / Each
Chemsteel S2106BCB-C1N47 PPS gear pump, 5.6 GPM, 115/230 VAC RK-70736-20 Mfr # OBS2106BCB-C1N47
COMPARE
100 5.6 PTFE-encapsulated silicone
$4,437.85CAD / Each
Chemsteel S2146BCB-C1T47 PPS gear pump, 8.0 GPM, 115/230 VAC RK-70736-25 Mfr # OBS2146BCB-C1T47
COMPARE
100 8 PTFE-encapsulated silicone
$4,120.34CAD / Each
Chemsteel S9176BCB-C1T47 PPS gear pump, 10 GPM, 115/230 VAC RK-70736-30 Mfr # OBS9176BCB-C1T47
COMPARE
100 10 PTFE-encapsulated silicone
$4,973.43CAD / Each
Chemsteel S9466BCB-C6X97 PPS gear pump, 30 GPM, 230/460 VAC RK-70736-50 Mfr # OBS9466BCB-C6X97
COMPARE
100 30 PTFE-encapsulated silicone
$6,492.49CAD / Each
Chemsteel R10216CA-F50 Stainless Steel Gear Pump; 0.5 GPM, 1/3 HP, 115/230 VAC RK-70736-60 Mfr # OBR10216CAC1-F50
COMPARE
110 0.58 PTFE-encapsulated silicone
$5,379.35CAD / Each
Chemsteel R10316CA-F50 316 Stainless Steel Gear Pump; 1.5 GPM, 115/230VAC RK-70736-62 Mfr # OBR10316CA-F50
COMPARE
150 1.5 PTFE-encapsulated silicone
$4,971.22CAD / Each
Chemsteel R10416CA-F50 316 Stainless Steel Gear Pump; 2 GPM, 115/230VAC RK-70736-64 Mfr # OBR10416CA-F50
COMPARE
150 2 PTFE-encapsulated silicone
$4,289.77CAD / Each
Chemsteel R10616CA-J47 316 Stainless Steel Gear Pump; 3 GPM, 115/230VAC RK-70736-66 Mfr # OBR10616CA-J47
COMPARE
150 3 PTFE-encapsulated silicone
$4,392.91CAD / Each
Chemsteel S20716CA-M30 316 Stainless Steel Gear Pump; 4 GPM, 115/230VAC RK-70736-68 Mfr # OBS20716CA-M47
COMPARE
150 4 PTFE-encapsulated silicone
$4,210.21CAD / Each
Chemsteel S20716CA-M30 316 Stainless Steel Gear Pump; 5.6 GPM, 115/230VAC RK-70736-70 Mfr # OBS20716CA-N47
COMPARE
150 5.6 PTFE-encapsulated silicone
$5,995.22CAD / Each
Chemsteel S21416CA-T97 316 Stainless Steel Gear Pump; 8 GPM, 115/230VAC RK-70736-72 Mfr # OBS21416CA-T97
COMPARE
150 8 PTFE-encapsulated silicone
$6,478.50CAD / Each
Chemsteel S41716CA-T97 316 Stainless Steel Gear Pump; 10 GPM, 115/230VAC RK-70736-74 Mfr # OBS41716CA-T97
COMPARE
150 10 PTFE-encapsulated silicone
$6,459.34CAD / Each
Chemsteel S92316CA-Y97 316 Stainless Steel Gear Pump; 15 GPM, 230/460 VAC RK-70736-76 Mfr # OBS92316CA-Y97
COMPARE
150 15 PTFE-encapsulated silicone
$7,666.05CAD / Each
Chemsteel S93016CA-Y97 316 Stainless Steel Gear Pump; 20 GPM, 230/460 VAC RK-70736-78 Mfr # OBS93016CA-Y97
COMPARE
150 20 PTFE-encapsulated silicone
$8,853.60CAD / Each
Chemsteel S93516CA-X97 316 Stainless Steel Gear Pump; 23 GPM, 230/460 VAC RK-70736-80 Mfr # OBS93516CA-X97
COMPARE
150 23 PTFE-encapsulated silicone
$8,339.39CAD / Each
Chemsteel S94616CA-X97 316 Stainless Steel Gear Pump; 30 GPM, 230/460 VAC RK-70736-82 Mfr # OBS94616CA-X97
COMPARE
150 30 PTFE-encapsulated silicone
$9,798.04CAD / Each
Showing 1- 20 of 20