CELLTREAT Scientific Products Oak Ridge Centrifuge Tubes
Ideal for high-speed, refrigerated, or standard centrifugation
  • Featuring leak-proof screw caps, uniform wall thickness, all-polypropylene construction, and round bottoms—these tubes can withstand up to 50,000 RCF x g
$92.82 - $135.55CAD / Case of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sterile Tube Bottom Shape Tube Capacity (mL)
Availability Pricing
CELLTREAT Scientific Products 229460 Oak Ridge Centrifuge Tube, 10 mL, Screw Cap, Bag, Nonsterile; 12/Cs RK-06336-25 Mfr # 229460
COMPARE
No Round/Autoclavable 10
$92.82CAD / Case of 12
CELLTREAT Scientific Products 229461 Oak Ridge Centrifuge Tube, 30 mL, Screw Cap, Bag, Nonsterile; 12/Cs RK-06336-26 Mfr # 229461
COMPARE
No Round/Autoclavable 30
$108.29CAD / Case of 12
CELLTREAT Scientific Products 229462 Oak Ridge Centrifuge Tube, 50 mL, Screw Cap, Bag, Nonsterile; 12/Cs RK-06336-27 Mfr # 229462
COMPARE
No Round/Autoclavable 50
$135.55CAD / Case of 12