CELLine Cell Culture Systems
Ideal for monoclonal antibody and cellular protein production—viable in vitro alternative to using animals
  • Separate growth and media chambers
The cell culture systems feature separate growth and media chambers which provide continuous supply of nutrients and direct oxygenation. The systems also eliminate waste buildup for high cell densities.
$576.10CAD / Case of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Celline CL1000 Cell Culture System, One 1000 mL Media Reservoir RK-13052-20 Mfr # WCL1000-1
COMPARE
Cell Culture System, One 1000 mL Media Reservoir
$576.10CAD / Case of 1