This product is now discontinued, View Related Products
CDI 4" Drive Digital Torque Tester, 5 to 50 in lb
Utilizes testing protocols and standards of the American Society of Mechanical Engineers (ASME)
  • Integral transducer and sturdy housing allow multiple mounting configurations
  • Full bridge strain gauge — 350 ohm, 1500 uE, 3 mV/V full scale, and 3.75 excitation
  • Audible and visible (LED) alert indicators
These economical digital torque testers are designed to be used on the bench or mounted on a wall allowing you to conveniently test torque wrenches and torque screwdrivers. Units feature an intuitive menu-driven, touch-pad screen that makes setup and operation easy. Testers display peak value readings in real time and provide the ability to download data to a PC for further analysis and report generation.
$5,159.81 - $6,049.00CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Capacity (N-m)
Availability Pricing
J. H. Williams Tool Group 1001-0-DTT Torque Tester, Digital, 10 to 100 In. Oz. RK-25301-30 Mfr # 1001-0-DTT
COMPARE
7.06
$5,833.15CAD / Each
J. H. Williams Tool Group 501-I-DTT Digital Torque Tester; 5 to 50 in lb RK-25301-31 Mfr # 501-I-DTT
COMPARE
5.65
$6,049.00CAD / Each
J. H. Williams Tool Group 4002-I-DTT Torque Tester, Digital, 40 to 400 In. lb. RK-25301-32 Mfr # 4002-I-DTT
COMPARE
45.2
$5,159.81CAD / Each
J. H. Williams Tool Group 2503-F-DTT Torque Tester, Digital, 25 to 250 ft. lb. RK-25301-33 Mfr # 2503-F-DTT
COMPARE
350
$5,423.55CAD / Each
J. H. Williams Tool Group 6004-F-DTT Torque Tester, Digital, 60 to 600 ft. lb. RK-25301-34 Mfr # 6004-F-DTT
COMPARE
813.5
$5,340.30CAD / Each