This product is now discontinued, View Related Products
Plas-Labs Catalyst Heaters
Reduces trace amounts of oxygen
Reduces trace amounts of oxygen and maintains incubation temperature levels of 35 to 37°C. Internal atmosphere is drawn up off floor and across heating elements and aluminized palladium pellets. Oxygen reacts with the anaerobic gas mixture and is reduced to water vapor. Measures 20"W x 12"H x 20"D.
$3,754.20CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC)
Availability Pricing
Plas-Labs 800-HEATER Catalyst Heater; 115 VAC RK-34749-56 Mfr # 800-HEATER
COMPARE
115
$3,754.20CAD / Each
Plas-Labs 800-HEATER/EXP Catalyst Heater; 220 VAC RK-34749-58 Mfr # 800-HEATER/EXP
COMPARE
220
$3,754.20CAD / Each