This product is now discontinued, View Related Products
Cascade Sciences Drying and Decarboxylation Ovens
Decrease run time and increase overall yield with more efficient horizontal airflow
  • Obtain a tight seal and excellent insulation from the snap-in fiberglass door gasket
  • Easily read temperature set point, timer, and humidity on digital display
  • Constructed with a durable stainless steel interior
  • Safely operate oven and protect samples with overtemperature shut-off
These dry and decarb forced-air ovens feature uniform air circulation and temperatures across the shelf space from right to left. This allows for optimal moisture removal and decarboxylation of plant material prior to further processing decreasing run time and increasing yields. These Cascade Sciences biomass ovens with built-in hygrometer are perfect for decarboxylation processes, drying organic matter, glass labware drying, and much more.
Large easy-to-view display with built-in timer and touch-button controls allows quick and simple temperature setting. Easily monitor the set temperature and moisture during the drying process to ensure optimal results. The exhaust port with sliding dampener doubles as an access port for external temperature sensors.
Choose from two oven options: the benchtop 4.9 cubic ft. model or the industrial 28 cubic ft. floor model. Both options feature a temperature range of ambient ±26° to 350°F. Typically decarboxylation practices require a temperature of 240°F for 1 hour and 15 minutes.
$14,800.20 - $18,742.99CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC) Rise Time
Availability Pricing
Cascade Sciences CDO-5 Drying and Decarboxylation Oven with Humidity Sensor, 4.9 Cubic Feet; 110-120 V, 50/60 Hz RK-42000-08 Mfr # CDO-5-CS
COMPARE
4.9 110 - 120 40 minutes to 240°F
$14,800.20CAD / Each
Cascade Sciences CDO-28 Drying and Decarboxylation Oven with Humidity Sensor, 28 Cubic Feet; 220 V, 50/60 Hz RK-42000-09 Mfr # CDO-28-CS
COMPARE
28 220 22 minutes to 240°F
$18,742.99CAD / Each