Corning® Pyrex® Caps and Pouring Rings for Bottles
Find replacements or upgrade your bottle's caps and pouring rings
  • Ensure the bottle cap size is the same as the bottle neck size
  • Polypropylene is autoclavable, PBT and ETFE can withstand dry heat sterilization
$41.25 - $302.04CAD / Case of 20
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Cap / Stopper Size Description
Availability Pricing
Corning Pyrex 1395-45LTC Polypropylene Caps for 1395 Media Bottles, Orange RK-34514-50 Mfr # 1395-45LTC
COMPARE
Yes GL45 Polypropylene Caps for Brand 1395 Media Bottles, orange
$145.87CAD / Case of 20
Corning Pyrex 1395-45LTC3 Polypropylene Caps for 1395 Media Bottles, Green RK-34514-53 Mfr # 1395-45LTC3
COMPARE
Yes GL45 Polypropylene Caps for Brand 1395 Media Bottles, green
$148.81CAD / Case of 20
Corning Pyrex 1395-45LTMC Polypropylene Caps for 1395 Media Bottles, Gray Membrane RK-34514-54 Mfr # 1395-45LTMC
COMPARE
Yes GL45 Polypropylene Caps for Brand 1395 Media Bottles, gray membrane
$117.87CAD / Case of 10
Corning Pyrex 1395-45HTC Polypropylene Caps for 1395 Media Bottles, High Temperature, Red RK-34514-55 Mfr # 1395-45HTC
COMPARE
Yes GL45 Polypropylene Caps for Brand 1395 Media Bottles, high temperature, red
$257.84CAD / Case of 10
Corning Pyrex 1395-45LTR Pour Rings for 1395 Media Bottles, Clear RK-34514-60 Mfr # 1395-45LTR
COMPARE
Yes GL45 Pour Rings for Brand 1395 Media Bottles, clear
$128.18CAD / Case of 50
Corning Pyrex 1395-45HTR Pour Rings for 1395 Media Bottles, High Temperature, Red RK-34514-65 Mfr # 1395-45HTR
COMPARE
Yes GL45 Pour Rings for Brand 1395 Media Bottles, high temperature, red
$302.04CAD / Case of 50
DWK Life Sciences (Kimble) 14395C-45 Replacement Cap for Media Bottles, PP, Blue; 10/CS RK-34523-11 Mfr # 14395C-45
COMPARE
Yes GL45 Replacement Cap for Media Bottles, PP, blue, 10/cs
$92.82CAD / Case of 10
DWK Life Sciences (Kimble) 14395P-45 Replacement Pouring Ring for Media Bottles, PP, Clear; 10/CS RK-34523-13 Mfr # 14395P-45
COMPARE
Yes GL45 Replacement Pouring Ring for Media Bottles, PP, clear, 10/cs
$41.25CAD / Case of 10
Showing 1- 8 of 8