InnoCal® Calibration Masses - ASTM Classes 0 to 3; OIML E2, F1, F2
ISO/IEC 17025:2005-Accredited Report
  • Provides traceability to NIST
  • Report lists the environmental conditions present at time of calibration
Mass calibrations are performed by an ISO 17025 accredited sub-contractor. InnoCal, our internal ISO 17025 accredited laboratory manages this sub-contracted service for our customers ensuring that the strictest quality requirements are being maintained. Calibration reports are issued directly to the customer after incoming inspection is performed by InnoCal's metrology staff. All measurements reported are traceable to SI units.
$102.84 - $795.63CAD / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
NIST Calibration Service for Masses. 26 to 30kg
$795.63CAD / Each
COMPARE
Recalibration report for calibration masses 1-25kg
$252.69CAD / Each
COMPARE
Recalibration report for calibration masses 101g-1kg
$194.49CAD / Each
COMPARE
NIST Calibration Service for Masses. 1 Mg to 100g; ASTM 0-3; Oiml E2, F1, F2
$102.84CAD / Each
COMPARE
NIST Calibration Service for Masses. >2 to 50lbs; ASTM 0-3; Oiml E2, F1, F2
$269.63CAD / Each
COMPARE
NIST Calibration Service for Masses. 0.51 to 2 Lbs; ASTM 0-3; Oiml E2, F1, F2
$194.49CAD / Each
COMPARE
NIST Calibration Service for Masses. 0.01 oz to 0.5 Lbs; ASTM 0-3; Oiml E2, F1, F2
$102.84CAD / Each
COMPARE
NIST Calibration Service for Masses. 2.1 oz to 8 oz; ASTM 0-3; Oiml E2, F1, F2
$179.02CAD / Each
COMPARE
NIST Calibration Service for Masses. 0.031 oz to 0.5 oz; ASTM 0-3; Oiml E2, F1, F2
$179.75CAD / Each
COMPARE
A2LA ISO17025 Accredited Recalibration Report for Calibration Masses 1 to 25 kg
$360.98CAD / Each
COMPARE
A2LA ISO17025 Accredited Recalibration Report for Calibration Masses 101 g to 1 kg
$302.04CAD / Each
COMPARE
A2LA ISO17025 Accredited Calibration; Calibration Weight/Mass, 1 mg to 100 g; ASTM 0-3; OIML E2, F1, F2
$210.69CAD / Each
COMPARE
A2LA ISO17025 Accredited Calibration; Calibration Weight/Mass, >2 to 50 lbs, ASTM 0-3; OIML E2, F1, F2
$377.19CAD / Each
COMPARE
A2LA ISO17025 Accredited Calibration; Calibration Weight/Mass, 0.51 to 2 lbs, ASTM 0-3; OIML E2, F1, F2
$302.04CAD / Each
COMPARE
A2LA ISO17025 Accredited Calibration; Calibration Weight/Mass, 0.01 oz to 0.5 lbs, ASTM 0-3; OIML E2, F1, F2
$210.69CAD / Each
COMPARE
A2LA ISO17025 Accredited Calibration; Calibration Weight/Mass, 2.1 oz to 8 oz; ASTM 0-3; OIML E2, F1, F2
$287.31CAD / Each
Showing 1- 16 of 16