This product is now discontinued, View Related Products
Cadence Science Micro-Emulsifying Needles
Quality and reliability for all your emulsifying needs
  • Luer lock tip for use with glass syringes
  • Available with or without reinforcing bar
Micro-Emulsifying needles are used in many applications including allergens. Mix two liquids by locking two syringes on the double hub needle and pump fliuids between the two syringes to produce a uniform mixture. Syringes are available with or without a reinforcing bar and in the overall lengths of 1-7/8 inches and 2-7/8 inches. Use with Luer Lock Tip glass syringes.
$169.44 - $775.00CAD / Pkg
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Needle Gauge
Availability Pricing
Cadence Micro-Emulsifying Needle with Reinforcing Bar, 25G 2-7/8" RK-25701-76 Mfr # 7974
COMPARE
25
$272.58CAD / Pkg
Cadence Micro-Emulsifying Needle with Reinforcing Bar, 22G 2-7/8" RK-25701-77 Mfr # 7975
COMPARE
22
$257.84CAD / Pkg
Cadence Micro-Emulsifying Needle with Reinforcing Bar, 20G 2-7/8" RK-25701-78 Mfr # 7976
COMPARE
20
$257.84CAD / Pkg
Cadence Micro-Emulsifying Needle with Reinforcing Bar, 18G 2-7/8" RK-25701-79 Mfr # 7977
COMPARE
18
$770.58CAD / Pkg of 4
Cadence Micro-Emulsifying Needle with Reinforcing Bar, 16G 2-7/8" RK-25701-80 Mfr # 7969
COMPARE
16
$775.00CAD / Pkg of 4
Cadence Micro-Emulsifying Needle with No Reinforcing Bar, 20G 1-7/8" RK-25701-88 Mfr # 7978
COMPARE
20
$198.91CAD / Pkg
Cadence Micro-Emulsifying Needle with No Reinforcing Bar, 18G 1-7/8" RK-25701-89 Mfr # 7979
COMPARE
18
$198.91CAD / Pkg
Cadence Micro-Emulsifying Needle with No Reinforcing Bar, 25G 2-7/8" RK-25701-96 Mfr # 7970
COMPARE
25
$169.44CAD / Pkg
Cadence Micro-Emulsifying Needle with No Reinforcing Bar, 22G 2-7/8" RK-25701-97 Mfr # 7971
COMPARE
22
$198.91CAD / Pkg
Cadence Micro-Emulsifying Needle with No Reinforcing Bar, 20G 2-7/8" RK-25701-98 Mfr # 7972
COMPARE
20
$198.91CAD / Pkg
Cadence Micro-Emulsifying Needle with No Reinforcing Bar, 18G 2-7/8" RK-25701-99 Mfr # 7973
COMPARE
18
$198.91CAD / Pkg
Showing 1- 11 of 11