This product is now discontinued, View Related Products
Cable Tester
Troubleshoot connectivity problems
  • Sees if your existing cabling can support voice, VoIP, 10/100/Gigabit Ethernet
  • Troubleshoots why existing cabling plant can’t support the desired network bandwidth
  • Discovers what’s at the end of any cable and displays device configuration (speed/duplex/pairing)
  • Frees-up switch ports and identifies unused links
  • Graphically maps wiring configuration and shows distance to faults with Intelligent Wiremap
  • Tests all copper cabling media: twisted-pair, coax, and audio cabling
  • Locates and traces cable with IntelliTone™ digital signaling technology
CableIQ is the first qualification tool designed for network technicians who need to troubleshoot connectivity problems and qualify cabling bandwidth. It will help you to determine if your cabling is qualifiedto support 10/100, VoIP or Gigabit Ethernet. Detects speed/duplex settings of attached switches and PCs. Intelligent wiremap graphically shows distance to faults. Intuitive, lightweight, and rugged.
$2,982.14 - $4,059.19CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Input Current (mA) Max Input Voltage (VDC) Number of Ports
Availability Pricing
Fluke Networks CIQ-KIT Networks CableIQ Advanced IT Kit RK-17782-51 Mfr # CIQ-KIT
COMPARE
$3,506.67CAD / Each
Fluke Networks CIQ-GSV 2788081 Cableiq Gigabit Service Kit RK-17791-63 Mfr # CIQ-GSV
COMPARE
$4,059.19CAD / Each
Fluke Networks CIQ-SVC 2577211 Networks CIQ-SVC CableIQ Service Kit RK-17791-65 Mfr # CIQ-SVC | 2577211
COMPARE
$3,403.53CAD / Each
Fluke Networks CIQ-100 CableIQ Qualification Tester RK-20021-84 Mfr # CIQ-100
COMPARE
$2,982.14CAD / Each