This product is now discontinued, View Related Products
Bud Industries UL/NEMA/IEC Plastic Enclosures
Gasketed enclosures can be used in a variety of wet, dirty, and corrosive environments
  • NEMA 4X, IEC529, IP65, and UL508-4X rated
ABS plastic enclosures are molded out of light gray material with UL94-HB flammability rating, and clear doors are polycarbonate plastic with UL94-V0 flammability rating. Plastic enclosures feature non-metallic hinges and snap-action closing latches. All three models have gasketed door for NEMA seal rating. Internal mounting bosses enable PCBs, process controls, and electrical junctions to be easily mounted within the enclosures. In order to maintain NEMA 4X rating, cable glands must be used when cables enter or exit the enclosure.
$64.39 - $338.88CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
Bud Industries NBB-10243 Plastic enclosure, type , solid door RK-94501-12 Mfr # NBB-10243
COMPARE
5.5 11.28 7.47
$116.40CAD / Each
Bud Industries NBB-10260 Plastic enclosure, type , clear door RK-94501-14 Mfr # NBB-10260
COMPARE
3.57 5.9 5.9
$64.39CAD / Each
Bud Industries NBB-10273 Plastic enclosure, type , clear door RK-94501-16 Mfr # NBB-10273
COMPARE
7.85 19.17 14.64
$338.88CAD / Each