This product is now discontinued, View Related Products
BriskHeat® Silicone Rubber Beaker Heater with X2 PID Temperature Controller Bundles
Ready-to-use heating cord bundles deliver complete systems for exceptional convenience
  • Programmable controller features alarm, user selectable °C or °F, and advanced PID control
  • Easy to read measurements with 0.4” high, 4-digit, green LED display
  • Designed with heating element laminated between two layers of 15 mil fiberglass reinforced silicone rubber
  • Maximum temperature of 232°C (450°F)
All-inclusive bundle features a digital PID controller with silicone rubber heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program, featuring a plug-and-play design. The X2 series controller offers a simple three button control with a programmable temperature, alarms, and security locks.
The flexible silicone rubber heating tape is moisture and chemical resistant to provide ideal heating for Griffin and standard size beakers. The heating tape is removable with hook and loop fastener for easy access to graduations.
Select heating tape length to optimize coverage while avoiding overlap.
$1,171.35 - $1,180.19CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Power (watts) Description
Availability Pricing
BriskHeat X2J-GBH0250-1 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 250 mL Beaker; 120 VAC RK-36120-95 Mfr # X2J-GBH0250-1
COMPARE
12 1/4 150 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 250 mL Beaker; 120 VAC
$1,171.35CAD / Each
BriskHeat X2J-GBH0400-1 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 400 mL Beaker; 120 VAC RK-36120-96 Mfr # X2J-GBH0400-1
COMPARE
13 3/4 200 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 400 mL Beaker; 120 VAC
$1,172.82CAD / Each
BriskHeat X2J-GBH0600-1 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 600 mL Beaker; 120 VAC RK-36120-97 Mfr # X2J-GBH0600-1
COMPARE
15 250 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 600 mL Beaker; 120 VAC
$1,174.29CAD / Each
BriskHeat X2J-GBH1000-1 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 1000 mL Beaker; 120 VAC RK-36120-98 Mfr # X2J-GBH1000-1
COMPARE
17 1/4 350 Silicone Rubber Griffin Beaker Heater with X2 PID Temperature Controller Bundle, for 1000 mL Beaker; 120 VAC
$1,180.19CAD / Each