This product is now discontinued, View Related Products
BriskHeat® Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle
Ready-to-use heating cord bundles deliver complete systems for exceptional convenience
  • Programmable controller features alarm, user-selectable °C or °F, and advanced PID control
  • Easy-to-read measurements with 0.4” high, 4-digit, green LED display
  • Type J thermocouple provides feedback for strict temperature control
  • Heating cord is constructed of double-braided high temperature Samox® fiberglass outer sheath with maximum temperature of 760°C (1400°F)
All-inclusive bundle features a digital PID controller with type J thermocouple, heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program, featuring a plug-and-play design. The X2 series controller offers a simple three-button control with a programmable temperature range of 0 to 760°C (32 to 1400°F). This advanced controller offers an accuracy of ±0.5°C (±0.9°F) with type J heating cords.
Select heating tape length to optimize coverage while avoiding overlap.
$1,163.98 - $1,234.70CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Max Temperature (° C) Description
Availability Pricing
BriskHeat X2J-HWC1040 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 4 ft length; 120 VAC RK-36120-91 Mfr # X2J-HWC1040
COMPARE
4 760 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 4 ft length; 120 VAC
$1,163.98CAD / Each
BriskHeat X2J-HWC1060 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 6 ft length; 120 VAC RK-36120-92 Mfr # X2J-HWC1060
COMPARE
6 760 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 6 ft length; 120 VAC
$1,180.19CAD / Each
BriskHeat X2J-HWC1120 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 12 ft length; 120 VAC RK-36120-93 Mfr # X2J-HWC1120
COMPARE
12 760 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 12 ft length; 120 VAC
$1,208.18CAD / Each
BriskHeat X2J-HWC1180 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 18 ft length; 120 VAC RK-36120-94 Mfr # X2J-HWC1180
COMPARE
18 760 Samox® Fiberglass High Temperature Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 18 ft length; 120 VAC
$1,234.70CAD / Each