This product is now discontinued, View Related Products
BriskHeat® Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundles
Ready-to-use heating cord bundles deliver complete systems for exceptional convenience
  • Programmable controller features alarm, user selectable °C or °F, and advanced PID control
  • Easy to read measurements with 0.4” high, 4-digit, green LED display
  • Type J thermocouple provides feedback for strict temperature control
  • Enhanced durability with heavy insulated design
  • Constructed of heavy insulated knitted and braided fiberglass
All-inclusive bundle features a digital PID controller with type J thermocouple, heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program, featuring a plug-and-play design. The X2 series controller offers a simple three button control with a programmable temperature range of 482°C (900°F). This advanced controller offers an accuracy with Type-J heating cords of ±0.5°C (±0.9°F).
The heavy insulated heating tapes feature an IP5X, rating making them ideal for rugged applications. Select heating tape length to optimize coverage while avoiding overlap.
$1,112.41 - $1,249.43CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Width (in) Description
Availability Pricing
BriskHeat X2J-BIH051020L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 2.0 ft; 120 VAC RK-36120-38 Mfr # X2J-BIH051020L
COMPARE
2 1/2 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 2.0 ft; 120 VAC
$1,112.41CAD / Each
BriskHeat X2J-BIH051040L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 4.0 ft; 120 VAC RK-36120-39 Mfr # X2J-BIH051040L
COMPARE
4 1/2 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 4.0 ft; 120 VAC
$1,128.62CAD / Each
BriskHeat X2J-BIH051060L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 6.0 ft; 120 VAC RK-36120-41 Mfr # X2J-BIH051060L
COMPARE
6 1/2 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 6.0 ft; 120 VAC
$1,153.66CAD / Each
BriskHeat X2J-BIH051080L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 8.0 ft; 120 VAC RK-36120-42 Mfr # X2J-BIH051080L
COMPARE
8 1/2 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 8.0 ft; 120 VAC
$1,181.66CAD / Each
BriskHeat X2J-BIH051100L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 10.0 ft; 120 VAC RK-36120-43 Mfr # X2J-BIH051100L
COMPARE
10 1/2 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 10.0 ft; 120 VAC
$1,214.07CAD / Each
BriskHeat X2J-BIH051120L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 12.0 ft; 120 VAC RK-36120-44 Mfr # X2J-BIH051120L
COMPARE
12 1/2 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 0.5 in x 12.0 ft; 120 VAC
$1,249.43CAD / Each
BriskHeat X2J-BIH101010L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 1.0 ft; 120 VAC RK-36120-46 Mfr # X2J-BIH101010L
COMPARE
1 1 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 1.0 ft; 120 VAC
$1,128.62CAD / Each
BriskHeat X2J-BIH101020L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 2.0 ft; 120 VAC RK-36120-47 Mfr # X2J-BIH101020L
COMPARE
2 1 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 2.0 ft; 120 VAC
$1,153.66CAD / Each
BriskHeat X2J-BIH101040L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 4.0 ft; 120 VAC RK-36120-48 Mfr # X2J-BIH101040L
COMPARE
4 1 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 4.0 ft; 120 VAC
$1,171.35CAD / Each
BriskHeat X2J-BIH101060L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 6.0 ft; 120 VAC RK-36120-49 Mfr # X2J-BIH101060L
COMPARE
6 1 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 6.0 ft; 120 VAC
$1,202.29CAD / Each
BriskHeat X2J-BIH101080L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 8.0 ft; 120 VAC RK-36120-50 Mfr # X2J-BIH101080L
COMPARE
8 1 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 8.0 ft; 120 VAC
$1,231.75CAD / Each
BriskHeat X2J-BIH101100L Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 10.0 ft; 120 VAC RK-36120-51 Mfr # X2J-BIH101100L
COMPARE
10 1 Heavy Insulated Fiberglass Heating Tape with X2 PID Temperature Controller Bundle, Type-J, 1.0 in x 10.0 ft; 120 VAC
$1,249.43CAD / Each
Showing 1- 12 of 12