This product is now discontinued, View Related Products
BriskHeat® Fiberglass Heating Cord with X2 PID Temperature Controller Bundles
Ready-to-use heating cord bundles deliver complete systems for exceptional convenience
  • Programmable controller features alarm, user-selectable °C or °F, and advanced PID control
  • Easy-to-read measurements with 0.4” high, 4-digit, green LED display
  • Type J thermocouple provides feedback for strict temperature control
  • Heating cord is constructed of double-braided fiberglass outer sheath with maximum temperature of 482°C (900°F)
All-inclusive bundle features a digital PID controller with type J thermocouple, heating tape, and power cords—everything you need to get started. Benchtop controller is compact and easy to program, featuring a plug-and-play design. The X2 series controller offers a simple three-button control with a programmable temperature range of 0 to 760°C (32 to 1400°F). This advanced controller offers an accuracy of ±0.5°C (±0.9°F) with type J heating cords.
Select heating tape length to optimize coverage while avoiding overlap.
$1,125.67 - $1,171.35CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Max Temperature (° C) Description
Availability Pricing
BriskHeat X2J-HTC451007 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 ft length; 120 VAC RK-36120-83 Mfr # X2J-HTC451007
COMPARE
0.5 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 0.5 ft length; 120 VAC
$1,125.67CAD / Each
BriskHeat X2J-HTC451006 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1 ft length; 120 VAC RK-36120-84 Mfr # X2J-HTC451006
COMPARE
1 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 1 ft length; 120 VAC
$1,127.14CAD / Each
BriskHeat X2J-HTC451005 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 2 ft length; 120 VAC RK-36120-85 Mfr # X2J-HTC451005
COMPARE
2 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 2 ft length; 120 VAC
$1,127.14CAD / Each
BriskHeat X2J-HTC451001 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 3 ft length; 120 VAC RK-36120-86 Mfr # X2J-HTC451001
COMPARE
3 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 3 ft length; 120 VAC
$1,130.09CAD / Each
BriskHeat X2J-HTC451008 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 4 ft length; 120 VAC RK-36120-87 Mfr # X2J-HTC451008
COMPARE
4 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 4 ft length; 120 VAC
$1,134.51CAD / Each
BriskHeat X2J-HTC451002 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 6 ft length; 120 VAC RK-36120-88 Mfr # X2J-HTC451002
COMPARE
6 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 6 ft length; 120 VAC
$1,144.82CAD / Each
BriskHeat X2J-HTC451009 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 8 ft length; 120 VAC RK-36120-89 Mfr # X2J-HTC451009
COMPARE
8 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 8 ft length; 120 VAC
$1,156.61CAD / Each
BriskHeat X2J-HTC451003 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 12 ft length; 120 VAC RK-36120-90 Mfr # X2J-HTC451003
COMPARE
12 482 Fiberglass Heating Cord with X2 PID Temperature Controller Bundle, Type-J, 12 ft length; 120 VAC
$1,171.35CAD / Each
Showing 1- 8 of 8