This product is now discontinued, View Related Products
BriskHeat® Benchtop Single Zone PID Temperature Controllers with Digital Display
Easy-to-use programmable digital controller delivers plug and play design
  • Programmable controller features alarm, user selectable °C or °F, and advanced PID control
  • Easy-to-read measurements with 0.4” high, 4-digit, green LED display
  • Enhanced safety with user programmable alarms including latching options
These X2-series digital benchtop temperature controllers provide advanced programming with PID control and connect easily to heating tape without additional wiring. The large display is easy-to-read and simple to program. Quickly program temperature, temperature units in °C or °F, alarm, security locks, timer, and much more.
Compatible with a wide range of heaters, these controllers include a factory installed Type-J or Type-K thermocouple sensor input. They are benchtop-friendly with a compact 6.39” x 9.48” x 1.99” profile.
Choose from models with or without stand clamp. Stand clamp allows controller to mount to poles with a 0.75” (18 mm) diameter or less. Heaters are sold separately.
$990.12CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
BriskHeat X2-120JTP Single Zone PID Temperature Controller, Fused Output, Type J Input; 120 VAC RK-36121-07 Mfr # X2-120JTP
COMPARE
1400 120 Single Zone PID Temperature Controller, Fused Output, Type J Input; 120 VAC
$990.12CAD / Each
BriskHeat X2-120KTP Single Zone PID Temperature Controller, Fused Output, Type K Input; 120 VAC RK-36121-08 Mfr # X2-120KTP
COMPARE
1400 120 Single Zone PID Temperature Controller, Fused Output, Type K Input; 120 VAC
$990.12CAD / Each
BriskHeat X2-120JSP Single Zone PID Temperature Controller with Stand Clamp, Fused Output, Type J Input; 120 VAC RK-36121-09 Mfr # X2-120JSP
COMPARE
1400 120 Single Zone PID Temperature Controller with Stand Clamp, Fused Output, Type J Input; 120 VAC
$990.12CAD / Each
BriskHeat X2-120KSP Single Zone PID Temperature Controller with Stand Clamp, Fused Output, Type K Input; 120 VAC RK-36121-10 Mfr # X2-120KSP
COMPARE
1400 120 Single Zone PID Temperature Controller with Stand Clamp, Fused Output, Type K Input; 120 VAC
$990.12CAD / Each