This product is now discontinued, View Related Products
BrandTech Safety-Coated Amber Glass Bottles
BrandTech Safety-Coated Amber Glass Bottles
  • Narrow-mouth bottles
  • Volumes of 250, 500, 1000, 2500 mL
  • Space-saving square base
  • Maximum operating temperature of 80°C
These amber glass bottles with square bases are coated with ethylene acrylate. Therefore, if the bottle breaks, its coating helps to reduce shattered glass. The bottles with polypropylene, screw caps and low-density polyethylene pouring rings are made of soda-lime glass. Cleaning temperatures should not exceed 60°C.
$41.62 - $115.66CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Bottle Color Capacity (mL) Cap Material
Availability Pricing
BrandTech 704004 Safety Bottle with Cap, Ethylene Acrylate-Coated Glass, Amber, 250 mL; 1/EA RK-06039-07 Mfr # 704004
COMPARE
Amber 250 polypropylene
$41.62CAD / Each
BrandTech 704006 Safety Bottle with Cap, Ethylene Acrylate-Coated Glass, Amber, 500 mL; 1/EA RK-06039-08 Mfr # 704006
COMPARE
Amber 500 polypropylene
$56.36CAD / Each
BrandTech 704008 Safety Bottle with Cap, Ethylene Acrylate-Coated Glass, Amber, 1000 mL; 1/EA RK-06039-09 Mfr # 704008
COMPARE
Amber 1000 polypropylene
$72.93CAD / Each
BrandTech 704010 Safety Bottle with Cap, Ethylene Acrylate-Coated Glass, Amber, 2500 mL; 1/EA RK-06039-10 Mfr # 704010
COMPARE
Amber 2500 polypropylene
$115.66CAD / Each