BRAND® HandyStep® Touch Pipettors
Next generation, touch screen repetitive pipette
  • Easy operation with context-sensitive help screens
The HandyStep® touch pipette is an electronic repeating pipettor with touch-screen operation, ability to store favorites, and motorized tip ejection to fit the needs of your application. It features an intuitive menu navigation, integrated help functions, and constantly displays the most important functions and parameters. The unique auto-dispense operating mode adapts to your dispensing rhythm without the need for programming. Motorized tip ejection by the press of a button prevents contamination and upholds sample integrity. The automatic tip size recognition of PD-Tip™ II pipette tips saves you time and prevents errors – simply insert the tip and the size is automatically recognized and displayed. When a new PD- Tip™ II tip of the same size is inserted, all settings are maintained. The USB-C connection allows for charging by cable; or inductively charge with the charging stand included in bundles.
$1,841.74 - $2,033.28CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (mL) Min Volume (µL) Description
Availability Pricing
BRAND HandyStep® Touch  Repeating Pipette Bundle RK-24806-01 Mfr # 705201
COMPARE
50 1 HandyStep® Touch Repeating Pipette Bundle
$2,033.28CAD / Each
BRAND HandyStep® Touch  Electronic Repeating Pipette, 100-240V RK-24806-04 Mfr # 705200
COMPARE
50 1 HandyStep® Touch Electronic Repeating Pipette, 100-240V
$1,841.74CAD / Each